Would you like to log in again?
Partisil® C8 5 and 10 micron HPLC Columns, USP-L7, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
4229-001 4239-001 4222-232
101946-518EA 1525.68 USD InStock
101946-518 101946-502 101946-494
Partisil® C8 5 and 10 micron HPLC Columns, USP-L7, Mac Mod
Chromatography Columns
Partisil C8 is an end-capped C8 monomeric phase with at least 8.5% carbon load
Partisil is a high-purity irregular silica gel available in both 5 µm and 10 µm particle sizes with a pore size of 80Å. The choice of column packing includes Silica, C-18 polymeric phases (ODS-3,ODS-2) and C-8. Also available are SAX, SCX, and PAC. These columns provide reproducible results, column to column, lot to lot.
Partisil C8 columns are available only in the fixed endfitting WCS hardware.
This column provides high efficiency and rapid mass transfer while maintaining excellent peak shape and stability over a range of aqueous mobile phase compositions. Recommended for ion pair chromatography.
Ordering information: Please see the Partisil/Partisphere accessories page for guard cartridges and hardware kits.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty