Cyanide Test Kit, Model CYN-3, Hach
Supplier: Hach
201002 201002
89230-922PK 258.44 USD InStock
CA11014-632 89230-922
Cyanide Test Kit, Model CYN-3, Hach
Environmental Test Kits
The main sources of cyanide, an extremely poisonous substance, are metal-cleaning and electroplating baths, gas scrubber, gas works, coke ovens, and various chemical processes.
  • Simple, lowcost method using a stepped color comparator
  • Achieve fast results with an easytocarry test kit
It can be removed easily from waste effluents through alkaline or neutral chlorination procedures. This kit provides a simple, low-cost method using a stepped color comparator.