Stablcal® Turbidity Standard, 1000 NTU, Hach
Supplier: Hach
Stablcal®
26606-49 2660649 2660642 2660642
97026-454EA 132.24 USD
CA26606-49 97026-454 97026-392 CA11016-420
Stablcal® Turbidity Standard, 1000 NTU, Hach
Stablcal® TuStablcal® Turbidity Standard for all Turbidimeters.
  • Precise, pre-mixed formazin solutions
  • No need for special glassware or difficult lab dilution techniques
  • Developed for use in any turbidimeter.
  • Enhanced stability saves time and labor
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter.