Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
FILLPALMINI
Supplier: Mettler Toledo
Peristaltic pump FillPalMini^ Peristaltic pump FillPalMini, 4 rolls peristaltic pump, SANTOPRENE peristaltic pump tube, 2.06 mm inner diameter (set of 2), includes tube set to connect to flow cell, can be operated in 2 pump directions at varying pump speeds, for sample recovery & cuvette cleaning.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILLPALMINI 10835-908 30254735 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty