KOVA REFRACTROL SP NORMAL 12X2ML
Supplier: Kova International
Refractol SP control^ Refractol SP control, Normal level, serum protein Solution for use in refractometry, for use with temperature compensated and non-compensated refractometers, provides assurance of measurements of plasma or serum protein concentration, Size: 12x2ml