Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
BACILLUS ATROPHAEUS SPORE STRIPS 10^4
Supplier: CROSSTEX
Spore Strip^ Bacillus atrophaeus Spore Strip (for Ethylene Oxide), Spore Population Per Strip: 10,000, monitoring various sterilization processes, Consist of inoculated filter paper, 6mm x 30mm, packaged in glassine pouches, Available in populations from 102 to 108 for each organism
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BACILLUS ATROPHAEUS SPORE STRIPS 10^4 10820-630 STN-04 Each (6Inch) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty