Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
20CF CORROSION PROOF EXP REF
Supplier: Thermo Scientific
Refrigerator^ Corrosion Resistant, Explosion-Proof Refrigerator, 20.0 cu. Ft. / 566.34L, 115V/60Hz, 4 fixed Interior shelves, 1 Wire Basket, 6 door shelves, Color White, Certifications/Compliance UL listed meets NFPA 45 and 70, 35.81 x 34 x 73.13 in, 5A, Door Style Hinged, Opens Left to Right
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
20CF CORROSION PROOF EXP REF 10818-364 20ERCETSA Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty