MR HEI-TEC 135MM PACKAGE
Supplier: HEIDOLPH NA LLC
Hotplate Package^ MR Hei-tec 135mm Magnetic Stirring Hotplate Package with PT1000 (Stainless Steel) Temperature Sensor, Rod, Clamp, Speed: 100 to 1,400 rpm (2% accuracy), Digital speed setting, Digital display, 115v