TIPPING STAND FOR LD25
Supplier: WORTHINGTON CRYOGENICS
Tipping stand^ Tipping stand,, allows safe dispensing of liquid nitrogen, supports the fully loaded weight, provides mobility, improves manueverability, has a 5-wheel cast-aluminum roller base, with side brackets, two locking wheels for pouring and additional handling ease