Shaking Incubator, Mini Shaker, SHEL LAB
Supplier: Sheldon Manufacturing
SSI2
89409-364EA 4293.71 USD InStock
89409-364
Shaking Incubator, Mini Shaker, SHEL LAB
Shakers and Mixers Shaking Incubators
The SHEL LAB Mini Shaker is the most compact shaking incubator in it's class
The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, petri dishes and staining trays.
Temperature, shaking speed and run time are displayed on the large LED control panel located on the front of the unit. The integral microprocessor includes a constant monitoring system that verifies and maintains accuracy through the duration of the program. Sophisticated over-temperature and over-speed controls ensure long life, safety and sample integrity.
Despite the remarkable small footprint, a large 11.5 x 9.5" work space accepts several platforms for use with a variety of flasks including 250ml and 500ml, tubes, and other common vessels.
Ordering information: 2-year limited warranty.