Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Model SD200 3-Channel Temperature Datalogger, Extech

Supplier: Extech
SD200 SD200-NIST
The 3-Channel Temperature Datalogger offers the ability to record data on an SD card in Excel® format

The SD200 features a triple LCD that simultaneously displays 3 Type-K temperature channels. Other features include a datalogger that stores readings on an SD card in Excel® format for easy transfer to a PC.

Certifications: CE marked.

Ordering information: Supplied complete with (6) AAA batteries, 2G SD card, univeral AC adaptor, (3) Type-K bead temperature probes, and mounting bracket. The unit also includes a manufacturer's one-year warranty.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
3-Channel Temperature Datalogger SD200 89200-738 Each Retrieving
3-Channel Temperature Datalogger with certificate of calibration traceable to NIST SD200-NIST 10171-764 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Resolution 0.1°
Basic Accuracy ±(0.5% + 0.5°C/1°F)
Type K Temperature –100 to 1300°C (–58 to 2372°F)

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty