Model RHT20 Humidity and Temperature Datalogger, Extech
Supplier: Extech
RHT20
97049-986EA 273.9 USD
97049-986
Model RHT20 Humidity and Temperature Datalogger, Extech
Data Loggers
Unit provides up to 16000 readings for relative humidity/temperature and user-programmable sample rate
  • Long battery life
  • Place or mount the datalogger in monitoring location (storage bin, truck, cargo hold, freezer, temperature-controlled room, and so on)
Model RHT20 features a selectable data sampling rate, as well as user-programmable alarm thresholds for relative humidity and temperature. The LCD displays current readings, min./max., and alarm status.
Certifications: CE marked.
Ordering information: Supplied complete with 3.6 V lithium battery, mounting bracket with combination lock, and Windows® 2000, XP, and Vista compatible analysis software. Unit also includes manufacturer's one-year warranty.