Model SD800 CO₂ / Humidity / Temperature Datalogger, Extech
Supplier: Extech
SD800
89200-744EA 849.54 USD
89200-744
Model SD800 CO₂ / Humidity / Temperature Datalogger, Extech
Data Loggers
The CO₂/Humidity/Temperature Datalogger records data on an SD card in Excel® format.
  • Maintenance free dual wavelength NDIR (non-dispersive infrared) CO₂ sensor
  • Checks for Carbon Dioxide (CO₂) concentrations
The SD500 features a triple LCD that simultaneously displays CO₂, temperature, and relative humidity. Other features include a datalogger that stores readings on an SD card in Excel® format for easy transfer to a PC.
It monitors air quality in schools, office buildings, greenhouses, hospitals and anywhere that high levels of carbon dioxide are generated.
Certifications: CE marked.
Ordering information: Supplied complete with (6) AAA batteries, 2G SD memory card, universal AC adaptor, and mounting bracket. The unit also includes a manufacturer's one-year warranty.