Model RH520A Humidity and Temperature Chart Recorder, Extech
Supplier: Extech
RH520A-NIST
10171-876EA 682.06 USD
10171-876
Model RH520A Humidity and Temperature Chart Recorder, Extech
Graphical datalogger for humidity/temperature measurements and dew point calculation
Model RH520A features a simultaneous numerical and graphical display of humidity and temperature readings, plus time and date. The unit also features internal memory with the capability to record up to 49,000 data points and can be transferred to a PC via RS-232 serial port for further data analysis.
Certifications: CE marked.
Ordering information: Supplied complete with built-in stand, detachable probe with 1m (3') cable, RS-232 cable, RS-232 to USB adaptor, LabView-based PC software, 100-240VAC 50/60Hz adaptor, and three AA batteries. Unit also includes manufacturer's one-year warranty.