HALO-5® PFP 5 micron Fused-Core® HPLC Column, USP-L43, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
95814509 95814309 95812409 95812509 95814709 95814409 95813409 95813509 95812909 95812309 95812209 95813309 95813709 95812609 95813909 95812709 95813609 95814909 95814609
102716-094EA 915.4 USD InStock
102716-094 102716-090 102716-068 102716-070 102716-098 102716-092 102716-080 102716-082 102716-076 102716-066 102716-064 102716-078 102716-086 102716-072 102716-088 102716-074 102716-084 102716-100 102716-096
HALO-5® PFP 5 micron Fused-Core® HPLC Column, USP-L43, Mac Mod
Chromatography Columns
Fused-Core particle technology was developed by Dr
On HALO-5 PFP, separations take place due to multiple types and degrees of analyte interactions including hydrophobic, pi–pi, dipole-dipole, and hydrogen bonding. Can be used in HILIC mode with mobile phases containing > 80% acetonitrile. Enhanced selectivity for stereoisomers. Compatible with highly aqueous mobile phases.
HALO PFP is available in 2.7 and 5 micron particle sizes. HALO PFP has a carbon load of 5.5% and a bonded phase coverage of 3.6 micromoles per square meter. HALO PFP has a pH range of 2 - 9 and a maximum operating temperature of 60 degrees C.
Jack Kirkland to produce UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower backpressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.
Ordering information: All 2.1, 3.0 and 4.6mm ID HALO HPLC and UHPLC columns use a cartridge guard system. Required for all guard cartridges is the HALO Guard Column Holder part number 94900-001 (See MAC-MOD Accessories page). The appropriate guard cartridge for a specific HALO column ID is listed under the Accessories Tab of the specific column page.