Pitot Tube Anemometer and Differential Manometer, Extech
Supplier: Extech
HD350
89200-746EA 937.83 USD
89200-746
Pitot Tube Anemometer and Differential Manometer, Extech
Anemometers
Model HD350 measures air velocity/airflow in difficult–to–reach or tight locations.
  • Simultaneous display for pressure, air velocity or air flow plus temperature.
  • Ability to store/recall up to 99 readings in each mode.
  • Features Max/Min/Avg recording, relative time stamp, data hold and auto power off function
Certifications: CE marked.
Ordering information: Three year warranty. Includes Windows® compatible software with cable, 9V battery, pitot tube with two 33.5” (85cm) connection hoses, and hard carrying case.