HALO® Phenyl Hexyl 2.7 micron Fused-Core® CapillaryUHPLC Column, USP-L11, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
98219406 98216406 98217706 98219706 92818706 92818406 98217406 98216706
101717-086EA 1242.86 USD InStock
101717-086 101717-070 101717-080 101717-088 101717-066 101717-064 101717-078 101717-076
HALO® Phenyl Hexyl 2.7 micron Fused-Core® CapillaryUHPLC Column, USP-L11, Mac Mod
Chromatography Columns
Fused-Core particle technology was developed by Dr
With HALO Phenyl-Hexyl, separations occur due to both hydrophobic and pi–pi interactions. Enhanced retention and selectivity for aromatic and unsaturated analytes (especially those with electron-withdrawing groups and halogens). Compatible with highly aqueous mobile phases.
HALO Phenyl-Hexyl is available in 2.7 and 5 micron particle sizes. HALO Phenyl-Hexyl has a carbon load of 7.1% and a bonded phase coverage of 3.6 micromoles per square meter. HALO Phenyl-Hexyl has a pH range of 2 - 9 and a maximum operating temperature of 60 degrees C.
Jack Kirkland to produce UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower backpressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.