INTERFACE I-100 - 11I100002
Supplier: BUCHI CORPORATION
Interface I-100^ Interface I-100, is an easy to use interface that controls the vacuum digitally. It is optimally suited to on/off-regulate the Vacuum Pump V-100, 160 x 105 x 120 mm, 100 - 240 V, For V-100 and V-700/V-710, with support set, communication cable, power pack