HALO® PFP 2.7 micron Fused-Core® UHPLC Column, USP-L43, Mac Mod
92812409 92813609 92814609 92813409 92812509 92813509 92813309 92814509 92812209 92814409 92814709 92812309 92813709 92814309 92812609
101668-512EA 960.06 USD
101668-512 101668-516 101668-526 101668-522 101668-514 101668-524 101668-520 101668-534 101668-508 101668-532 101668-528 101668-510 101668-518 101668-530 101668-504
HALO® PFP 2.7 micron Fused-Core® UHPLC Column, USP-L43, Mac Mod
Chromatography Columns
Fused-Core particle technology was developed by Dr
With HALO PFP, separations take place due to multiple types and degrees of analyte interactions including hydrophobic, pi–pi, dipole-dipole, and hydrogen bonding. Can be used in HILIC mode with mobile phases containing > 80% acetonitrile. Enhanced selectivity for stereoisomers. Compatible with highly aqueous mobile phases.
HALO PFP is available in 2.7 and 5 micron particle sizes. HALO PFP has a carbon load of 5.5% and a bonded phase coverage of 3.6 micromoles per square meter. HALO PFP has a pH range of 2 - 9 and a maximum operating temperature of 60 degrees C.
Jack Kirkland to produce UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower backpressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.
Ordering information: All 2.1, 3.0 and 4.6mm ID HALO HPLC and UHPLC columns use a cartridge guard system. Required for all guard cartridges is the HALO Guard Column Holder part number 94900-001 (See MAC-MOD Accessories page). The appropriate guard cartridge for a specific HALO column ID is listed under the Accessories Tab of the specific column page.