HALO® HILIC 2.7 micron Fused-Core® UHPLC Column, USP-L3, Mac Mod
92813501 92814201 92814501 92814401 92814301 92814701 92812701 92812401 92813701 92813601 92813201 92812501 92813301 92812301 92812201 92814601 92813401 92812601
101668-490EA 1004.71 USD
101668-490 101668-496 101668-502 101668-500 101668-498 101668-494 101668-470 101668-476 101668-482 101668-480 101668-484 101668-478 101668-486 101668-474 101668-472 101668-492 101668-488 101668-468
HALO® HILIC 2.7 micron Fused-Core® UHPLC Column, USP-L3, Mac Mod
Chromatography Columns
Fused-Core particle technology was developed by Dr
HALO HILIC can be used in both HILIC and normal-phase modes. Enhanced sensitivity and peak shape for LC-MS analyses of basic analytes.
HALO HILIC is available in 2.7 and 5 micron particle sizes. HALO HILIC has a surface area of 150 square meters per gram. There is no bonded phase on the silica surface. HALO HILIC has a pH range of 1 - 8 and a maximum operating temperature of 60 degrees C.
Jack Kirkland to produce UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower backpressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.
Ordering information: All 2.1, 3.0 and 4.6mm ID HALO HPLC and UHPLC columns us a cartridge guard system. Required for all guard cartridges is the HALO Guard Column Holder part number 94900-001 (See MAC-MOD Accessories page). The appropriate guard cartridge for a specific HALO column ID is listed under the Accessories Tab of the specific column page.