Model CO200 Desktop Indoor Air Quality CO₂ Monitor, Extech
Supplier: Extech
CO200
97050-002EA 407.4 USD
97050-002
Model CO200 Desktop Indoor Air Quality CO₂ Monitor, Extech
The Desktop Indoor Air Quality CO₂ Monitor measures carbon dioxide (CO₂), air temperature, and humidity
Model CO200 checks for carbon dioxide concentrations with a min./max. value recall function. It features a visible and audible CO₂ warning alarm with relay output for ventilation control, as well as displaying temperature, humidity, year, month, date, and time.
Certifications: CE marked.
Ordering information: Supplied complete with universal AC adaptor and manufacturer's one-year warranty.