Would you like to log in again?
Model CO10 Carbon Monoxide Meter, Extech
Supplier: Extech
CO10
89370-182EA 377.78 USD InStock
89370-182
Model CO10 Carbon Monoxide Meter, Extech
Gas Detection Systems
The Carbon Monoxide Meter offers faster response measurements up to 1000ppm
The model CO10 meter features an easy to read 1999 count backlit display with CO levels from 0 to 1000ppm. It utilizes a stabilized electrochemical gas specific (CO) fast response sensor. Other features include audible alarm and backlight for low light conditions.
Certifications: CE marked.
Ordering information: Supplied complete with 9V battery, protective rubber holster, and carrying case. Unit also includes manufacturer's one-year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty