HALO® Peptide ES CN Fused-Core® UHPLC Column, USP-L10, 2.7 μm, Mac Mod
92812704 92813504 92812304 92813704 92813304 92813404 92814304 92814604 92812604 92812404 92814404 92812204 92813604 92814504 92814704 92812504
101717-032EA 840.98 USD
101717-032 101717-054 101717-042 101717-056 101717-044 101717-022 101717-060 101717-052 101717-040 101717-018 101717-036 101717-058 101717-014 101717-024 101717-016 101717-026
HALO® Peptide ES CN Fused-Core® UHPLC Column, USP-L10, 2.7 μm, Mac Mod
Fused-Core particle technology was developed by Dr.
HALO Peptide ES CN gives complementary selectivity to other HALO C18 and C8, Phenyl-Hexyl, RP-Amide, and PFP phases. Enhanced stability at low pH up to 90 °C via sterically protected ligands. Ideal for reducing hydrophobic analyte retention for samples containing both nonpolar and polar analytes.
HALO ES-CN is available in 2.7 and 5 µm particle sizes. HALO ES-CN has a carbon load of 3.5% and a bonded phase coverage of 2.5 micromoles per square meter. HALO ES-CN has a pH range of 1 to 9 and a maximum operating temperature of 90 °C.
Jack Kirkland to produce UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower backpressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.
Ordering information: All 2.1, 3.0 and 4.6 mm Int.Ø HALO HPLC and UHPLC columns use a cartridge guard system. Required for all guard cartridges is the HALO Guard Column Holder part number 94900-001 (See MAC-MOD Accessories page). The appropriate guard cartridge for a specific HALO column Int.Ø is listed under the Accessories Tab of the specific column page.