HALO® C18 2.7 micron Fused-Core® UHPLC Column, USP-L1, Mac Mod
92811602 92814702 92813402 92812702 92812402 92814602 92814202 92813502 92813602 92812502 92812602 92814502 92814302 92811702 92812202 92811402 92810702 92814402 92813302 92813702 92813202 92811302 92812302
101717-006EA 1101.46 USD
101717-006 101668-390 101668-384 101668-366 101668-372 101668-388 101668-392 101668-386 101668-376 101668-374 101668-364 101668-398 101668-394 101717-010 101668-368 101717-000 101716-992 101668-396 101668-382 101668-378 101668-380 101716-996 101668-370
HALO® C18 2.7 micron Fused-Core® UHPLC Column, USP-L1, Mac Mod
Chromatography Columns
HALO C18 gives excellent performance for a broad range of analyte polarities
HALO C18 gives excellent performance for a broad range of analyte polarities. Separations are due primarily to hydrophobic interactions and differences in hydrophobicity among analytes.
  • Ultra-high resolution and ultra-fast separations without sacrificing column ruggedness and reliability
  • Used with most conventional HPLC equipment for UHPLC performance
  • Easy transfer of methods from UHPLC to HPLC
Fused-Core particle technology was developed by Dr. Jack Kirkland to produce UHPLC columns that provide ultra-high resolution and ultra-fast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower back pressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.
Separations are due primarily to hydrophobic interactions and differences in hydrophobicity among analytes.
Ordering information: All 2.1, 3.0 and 4.6mm ID HALO HPLC and UHPLC columns use a cartridge guard system. Required for all guard cartridges is the HALO Guard Column Holder part number 94900-001 (See MAC-MOD Accessories page). The appropriate guard cartridge for a specific HALO column ID is listed under the Accessories Tab of the specific column page.