HALO® C8 Fused-Core® UHPLC Column, USP-L7, 2.7 μm, Mac Mod
92812608 92814408 92813208 92812708 92812508 92813508 92814308 92810708 92812408 92812208 92814208 92812308 92813408 92814608 92814508 92813608 92813308 92813708 92814708
101668-418EA 1213.09 USD
101668-418 101668-448 101668-426 101668-436 101716-994 101668-428 101668-416 101668-438 101668-444 101668-422 101668-432 101668-446 101668-424 101668-434 101668-440 101668-450 101668-420 101668-442 101668-430
HALO® C8 Fused-Core® UHPLC Column, USP-L7, 2.7 μm, Mac Mod
Fused-Core particle technology was developed by Dr.
HALO C8 gives excellent performance for a broad range of analyte polarities. Separations are due primarily to hydrophobic interactions and differences in hydrophobicity among analytes.
Jack Kirkland to produce UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower backpressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.
Ordering information: All 2.1, 3.0 and 4.6 mm Int.Ø HALO HPLC and UHPLC columns use a cartridge guard system. Required for all guard cartridges is the HALO Guard Column Holder part number 94900-001 (See MAC-MOD Accessories page). The appropriate guard cartridge for a specific HALO column Int.Ø is listed under the Accessories Tab of the specific column page.