Would you like to log in again?
HALO® RP Amide 2.7 micron Fused-Core® UHPLC Column, USP-L60, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
92813607 92814407 92812407 92813507 92813707 92814507 92813407 92812707 92812607 92814607 92814207 92812307 92814307 92813307 92812507 92814707 92812207 92813207
101668-610EA 1131.23 USD InStock
101668-610 101668-616 101717-020 101668-608 101717-046 101668-618 101717-038 101668-602 101668-600 101668-612 101668-626 101668-604 101668-614 101717-034 101668-606 101717-062 101668-620 101668-624
HALO® RP Amide 2.7 micron Fused-Core® UHPLC Column, USP-L60, Mac Mod
Chromatography Columns
Fused-Core particle technology was developed by Dr
With HALO RP Amide, retention and selectivity are due to both hydrophobic and hydrogen bonding interactions. Less retentive than C18 and C8, except for analytes with proton-donating groups (alcohols, acids, etc.). Compatible with 100% aqueous mobile phases.
HALO RP-Amide is available in a 2.7 micron particle size. HALO RP-Amide has a carbon load of 8.2% and a bonded phase coverage of 3.0 micromoles per square meter. HALO RP-Amide has a pH range of 2 - 9 and a maximum operating temperature of 60 degrees C.
Jack Kirkland to produce UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability. HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. It is this moderate back pressure of HALO columns that also permits them to be used with most conventional HPLC equipment for UHPLC performance. The combination of much lower back pressure and excellent efficiency at higher linear velocities permits the easy transfer of methods from UHPLC to HPLC, and vice versa.
Ordering information: All 2.1, 3.0 and 4.6mm ID HALO HPLC and UHPLC columns use a cartridge guard system. Required for all guard cartridges is the HALO Guard Column Holder part number 94900-001 (See MAC-MOD Accessories page). The appropriate guard cartridge for a specific HALO column ID is listed under the Accessories Tab of the specific column page.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty