Hardness and Iron Color Disc Test Kit, Model HA-77, Hach
Supplier: Hach
202300
89231-260PK 143.43 USD InStock
89231-260
Hardness and Iron Color Disc Test Kit, Model HA-77, Hach
Environmental Test Kits
Hardness & Iron Water Quality Test Kit is the kit to use if you need better accuracy than that provided by a test cube.
  • The kit to use for more resolution in a visual test
  • Uses simple, economical drop count titration and color disc methods
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Utilizing drop count titration and color disc colorimetry, this kit contains 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.