2,2,2-Trichloro-1-(1-methyl-4-nitro-1H-pyrrol-2-yl)ethanone
Synonyms:
1-Methyl-4-nitro-2-(trichloroacetyl)-1H-pyrrole
4027090.0001
4027090.0005
Q-2650.0001BAEA
371.47
USD
Q-2650.0001BA
Q-2650.0005BA
2,2,2-Trichloro-1-(1-methyl-4-nitro-1H-pyrrol-2-yl)ethanone
2,2,2-Trichloro-1-(1-methyl-4-nitro-1H-pyrrol-2-yl)ethanone
Sequence: 1-Methyl-4-nitro-2-(trichloroacetyl)-1H-pyrrole
Synonym(s): 2,2,2-Trichloro-1-(1-methyl-4-nitro-1H-pyrrol-2-yl)-ethanone
Building block for the synthesis of DNA-binding molecules, such as the minor groove-binding antitumor agents distamycin A and netropsin.
Formula:
C₇H₅Cl₃N₂O₃ Storage Temperature: Freezer |
MDL Number:
MFCD03789117 CAS Number: 120122-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...