Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Fmoc-hydrazino)benzoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: 4-(Fmoc-hydrazino)-benzoic acid

Q-2545.0001 Q-2545.0005
Q-2545.0001BAEA 83.64 USD InStock
Q-2545.0001BA Q-2545.0005BA
4-(2-Fmoc-hydrazino)benzoic acid
4-(2-Fmoc-hydrazino)benzoic acid
Sequence: 4-(Fmoc-hydrazino)-benzoic acid
CAS-No.: 214475-53-3
Mol.weight: 374.40
SumFormula: C₂₂H₁₈N₂O₄
Bulk item no.: Q-2545
Synonym(s):
EINECS no.: -
MDL no.: MFCD02179643

This linker which is stable to acid and bases is cleaved with high specificity under mild oxidative conditions in the presence of base and a nucleophile, yielding a range of C-terminal functionalities. The linker may be used to synthesize fully or partially protected peptides for fragment coupling. See also D-2560.


Formula: C₂₂H₁₈N₂O₄
MW: 374,396 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179643
CAS Number: 214475-53-3
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty