Would you like to log in again?
N-Boc-ethylenediamine
Supplier: BACHEM AMERICAS INC
Synonyms: t-boc-N-EDAN-BOC-1,2-diaminoethane
Q-2450.0005 Q-2450.0001
Q-2450.0005BAEA 334.2 USD InStock
Q-2450.0005BA Q-2450.0001BA
N-Boc-ethylenediamine
N-Boc-ethylenediamine
Sequence: N-1-Boc-1,2-diaminoethane · HCl
Mono-protected alkylene diamines are important homobifunctional compounds often used as spacer groups (especially hexamethylenediamine), e.g. for the immobilization of peptides. Additionally they are very useful building blocks for preparing polyamines and polyamides. 1,5-Diaminopentane (cadaverine) is the decarboxylation product of lysine, ornithine yields 1,4-diaminobutane (putrescine).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty