4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid

Synonyms: 4-(4-(1-Hydroxyethyl)-2-methoxy-5-nitro-phenoxy)butyric acid

4027672.0250 4027672.1000
Q-2345.0250BAEA 371.47 USD
Q-2345.0250BA Q-2345.1000BA
4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid
4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid

Sequence: 4-(4-(1-Hydroxyethyl)-2-methoxy-5-nitro-phenoxy)butyric acid


The photolabile linker 4-[4-(1-hydroxyethyl)-2-methoxy-5-nitrophenoxy]butanoic acid ("hydroxyethyl photolinker") is a valuable tool for the preparation of combinatorial libraries, as it allows release from the solid support under neutral conditions to give libraries of acids.

Formula: C₁₃H₁₇NO₇
Storage Temperature: Refrigerator
MDL Number: MFCD01318869
CAS Number: 175281-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR