Pmc-S-methylisothiourea

Supplier: BACHEM AMERICAS INC

4027659.0001
Q-2330.0001BAEA 152.38 USD
Q-2330.0001BA
Pmc-S-methylisothiourea
Pmc-S-methylisothiourea

Sequence: Pmc-S-methylisothiourea


For DOTA(OtBu)₃ see Q-2525.


Reagent for guanylation of primary, secondary and aryl amines. The Pmc group is stable to most organometallics, base, hydrogenolysis, but requires more vigorous conditions for removal, thus providing orthogonal protection to standard Z, Boc and Fmoc protection strategies.

Formula: C₁₆H₂₄N₂O₃S₂
Storage Temperature: Refrigerator
MDL Number: MFCD01318867
CAS Number: 185674-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR