3-(4-Azidophenyl)propionic acid

Supplier: BACHEM AMERICAS INC

Q-1995.0001 Q-1995.0005
Q-1995.0001BAEA 313.7 USD
Q-1995.0001BA Q-1995.0005BA
3-(4-Azidophenyl)propionic acid
3-(4-Azidophenyl)propionic acid

Sequence: 3-(4-Azidophenyl)propionic acid
CAS-No.: 103489-31-2
Mol.weight: 191.19
SumFormula: C₉H₉N₃O₂
Bulk item no.: Q-1995
Synonym(s):
EINECS no.: -
MDL no.: MFCD00798799


Buiding blocks for "click chemistry". See also the product family "Propargylglycine" and the subfamily "p-Azidophenylalanine". Bachem additionally offers azido derivatives for SPPS: Fmoc-Dab(N₂)-OH (B-4145), Fmoc-Orn(N₂)-OH (B-4140), Fmoc-Lys(N₂)-OH (B-4135), Boc-Orn(N₂)-OH · DCHA (A-4770), and Boc-Lys(N₂)-OH · DCHA (A-4765).

Formula: C₉H₉N₃O₂
MW: 191,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00798799
CAS Number: 103489-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR