(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)

Synonyms: 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphateBenzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V)BOP reagent

4031234.0100 4031234.0025
Q-1980.0100BAEA 718.66 USD
Q-1980.0100BA Q-1980.0025BA
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)

Sequence: Benzotriazol-1-yloxy-tris(dimethylamino)-phosphonium hexafluorophosphate


Coupling reagent for peptide synthesis, which suppresses enantiomerization. Useful reagent for lactonization, selective esterification or amidation of α-amino acids without racemization.

Formula: C₁₂H₂₂F₆N₆OP₂
MW: 442.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011948
CAS Number: 56602-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR