BACEQ-1645
4015245.0005 4015245.0025 4015245.0001
Q-1645.0025BAEA 1708.76 USD
Q-1645.0025BA Q-1645.0001BA Q-1645.0005BA
BACEQ-1645
Sequence: (2-Methylindol-1-yl)acetic acid · DCHA
For DOTA(OtBu)₃ see Q-2525.
Scavenger for the removal of the Nα-protecting group o-nitrophenylsulfenyl (Nps) group with ammonium thiocyanate.