Melatonine

Synonyms: MelatoninN-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

4008335.0005
Q-1300.0005BAEA 136.9 USD
Q-1300.0005BA
Melatonine
Melatonine

Sequence: N-Acetyl-5-methoxytryptamine


For luzindole see Q-1885.


Melatonin reverses the darkening effect (on skin) of melanotropin and inhibits LH-secretion. Also available in bulk quantities.

Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005655
CAS Number: 73-31-4
EINECS: 200-797-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR