BACEQ-1120
4028760.0005 4028760.0001
Q-1120.0005BAEA 346.7 USD
Q-1120.0005BA Q-1120.0001BA
BACEQ-1120
Sequence: 5-Bromo-7-nitroindoline
A reagent for introducing a protecting group for carboxylic acids (including amino acids and peptides) that can be photosolvolyzed to give acids, esters or amides (peptides).