N-Dodecanoyl-N-methyl-D-glucamine (MEGA-12)

P-1175.0005 P-1175.0001
P-1175.0005BAEA 385.8 USD
P-1175.0005BA P-1175.0001BA
N-Dodecanoyl-N-methyl-D-glucamine (MEGA-12)
N-Dodecanoyl-N-methyl-D-glucamine (MEGA-12)

Sequence: MEGA-12
Synonym(s): Dodecanoyl-N-methylglucamide#N-Dodecanoyl-N-methyl-D-glucamine


Non-ionic detergents with high solubilizing power and nondenaturing properties. They can easily be removed by dialysis. Bachem offers MEGA-8, -9, -10, and -12.

Formula: C₁₉H₃₇NO₇
Storage Temperature: Freezer
CAS Number: 87246-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR