Nonyl-β-D-glucopyranoside

Synonyms: β-Nonylglucoside(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(nonyloxy)tetrahydro-2H-pyran-3,4,5-triol

4026563.0005 4026563.0001
P-1150.0001BAEA 141.13 USD
P-1150.0001BA P-1150.0005BA
Nonyl-β-D-glucopyranoside
Nonyl β-D-glucopyranoside

Sequence: n-Nonyl β-D-glucopyranoside
Synonym(s): n-Nonyl β-D-glucoside-(1,5)#NGP


NGP, non-ionic detergent with a CMC of 6.5 mM has been shown to be suitable for membrane research.

Formula: C₁₅H₃₀O₆
MW: 306.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063300
CAS Number: 69984-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR