Oleamide

Supplier: BACHEM AMERICAS INC

Synonyms: cis-9-OctadecenamideOleyramide

O-1570.0500 4026424.0100
O-1570.0500BAEA 860.58 USD InStock
O-1570.0500BA O-1570.0100BA
Oleamide
Oleamide

Sequence: cis-9,10-Octadecenamide
CAS-No.: 301-02-0
Mol.weight: 281.48
SumFormula: C₁₈H₃₅NO
Bulk item no.: O-1570
Synonym(s): Oleamide
EINECS no.: 206-103-9
MDL no.: MFCD00053638


Oleamide, a mono-unsaturated fatty acid amide, was isolated from the cerebrospinal fluid of sleep-deprived cats. Synthetic oleamide induced physiological sleep when injected into rats.

Formula: C₁₈H₃₅NO
MW: 281.48 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00053638
CAS Number: 301-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR