BACEO-1015

4004273.0005
O-1015.0005BAEA 742.94 USD
O-1015.0005BA
BACEO-1015
3-O-Benzyl-sn-glycerol
CAS: 56552-80-8
Formula: C10H14O3
M.W: 182.22
Storage temp.: -20 ± 5 °C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR