Biotinyl-Val-Ala-DL-Asp-fluoromethylketone
Supplier: Bachem
4034787.0001
N-1840.0001BAEA 1004.06 USD
N-1840.0001BA
Biotinyl-Val-Ala-DL-Asp-fluoromethylketone
Proteins and Peptides
See also the Asp derivatives N-1425 and N-1430, and Z-YVAD-FMK (N-1595).
Biotinylated analog of the caspase-1 inhibitor Z-VAD-FMK, N-1510.