Ac-D-d-Gla-L-I-Cha-C-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Asp-D-Gla-Leu-Ile-β-cyclohexyl-Ala-Cys-OH

4030703.0001 4030703.0005
N-1725.0001BAEA 835.51 USD
N-1725.0001BA N-1725.0005BA
Ac-D-d-Gla-L-I-Cha-C-OH
Ac-D-d-Gla-L-I-Cha-C-OH

Sequence: Ac-Asp-D-Gla-Leu-Ile-β-cyclohexyl-Ala-Cys-OH
CAS-No.: 208940-40-3
Mol.weight: 830.95
SumFormula: C₃₆H₅₈N₆O₁₄S
Bulk item no.: N-1725
Synonym(s): Ac-Asp-DGla-Leu-Ile-b-cyclohexyl-Ala-Cys
EINECS no.: -
MDL no.: MFCD02259688


Ac-Asp-D-Gla-Leu-Ile-Cha-Cys is a potent inhibitor of HCV NS3 protease with IC₅₀ values of 1.5 nM (in the absence of NaCl) and 40 nM (in the presence of 150 mM NaCl).

Formula: C₃₆H₅₈N₆O₁₄S
MW: 830.948 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02259688
CAS Number: 208940-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR