Ac-1-Nal-Abu-Phe-psi(CH₂NH)Abu-Abu-1-Nal-NH₂

Supplier: BACHEM AMERICAS INC

4030621.0001 4030621.0005
N-1705.0001BAEA 645.34 USD InStock
N-1705.0001BA N-1705.0005BA
Ac-1-Nal-Abu-Phe-psi(CH₂NH)Abu-Abu-1-Nal-NH₂
Proteins and Peptides
Sequence: Ac-1-Nal-Abu-Phe-psi(CH₂NH)Abu-Abu-1-Nal-NH₂
CAS-No.: 240811-12-5
Mol.weight: 842.05
SumFormula: C₄₉H₅₉N₇O₆
Bulk item no.: N-1705
Synonym(s):
EINECS no.: -
MDL no.: MFCD02259685

Ac-1-Nal-Abu-Phe-psi(CH₂NH)-Abu-Abu-1-Nal-NH₂, a designed inhibitor for feline immunodeficiency virus (FIV) protease (Ki = 20 nM).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR