H-D-Phe-Pro-Arg-chloromethylketone Trifluoroacetate
Supplier: Bachem
4033019.0025 4033019.0100
N-1065.0025BAEA 2189.75 USD
N-1065.0025BA N-1065.0100BA
H-D-Phe-Pro-Arg-chloromethylketone Trifluoroacetate
Proteins and Peptides
See also lepirudin (H-6618) and bivalirudin (H-6668).
PPACK, fPR-CMK, a highly effective irreversible inhibitor of thrombin and gingipain R. For biotinylated PPACK see N-1565.