Z-Phe-Ala-diazomethylketone

Supplier: BACHEM AMERICAS INC

4003801.0050 4003801.0250
N-1040.0050BAEA 428.2 USD
N-1040.0050BA N-1040.0250BA
Z-Phe-Ala-diazomethylketone
Z-Phe-Ala-diazomethylketone

Sequence: Z-Phe-Ala-diazomethylketone
CAS-No.: 71732-53-1
Mol.weight: 394.43
SumFormula: C₂₁H₂₂N₄O₄
Bulk item no.: N-1040
Synonym(s):
EINECS no.: -
MDL no.: MFCD00077029


Z-FA-CHN2, a specific inhibitor of thiol proteinases, e.g. cathepsin B. The diazomethyl ketones Z-FA-CHN2 and Z-LVG-CHN2 (C-3455) inhibited cathepsin L-like cysteine proteinase B from Leishmania mexicana.

Formula: C₂₁H₂₂N₄O₄
MW: 394.431 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077029
CAS Number: 71732-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR