Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

FA-Glu-Glu-OH

Supplier: BACHEM AMERICAS INC

M-2385.0250 M-2385.0050
M-2385.0250BAEA 2149.67 USD InStock
M-2385.0250BA M-2385.0050BA
FA-Glu-Glu-OH
Proteins and Peptides
Sequence: FA-Glu-Glu-OH
CAS-No.: 1374423-90-1
Mol.weight: 396.35
SumFormula: C₁₇H₂₀N₂O₉
Bulk item no.: M-2385
Synonym(s):
EINECS no.: -
MDL no.: MFCD03093409

The FA-peptides M-1400 and M-1410 are common ACE substrates and thus filed in the corresponding product family. For our 3-(2-furyl)acryloyl-amino acids (FA-amino acids) and the labeling reagent FA-OSu (Q-1245), please see the Amino Acids section.

Preferred substrate of human and zebrafish carboxypeptidase O.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty