Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ac-Asp-Glu-Asp(EDANS)-Glu-Glu-Abu-L-lactoyl-Ser-Lys(DABCYL)-NH₂

Supplier: BACHEM AMERICAS INC

M-2235.1000 M-2235.0500
M-2235.1000BAEA 656.47 USD InStock
M-2235.1000BA M-2235.0500BA
Ac-Asp-Glu-Asp(EDANS)-Glu-Glu-Abu-L-lactoyl-Ser-Lys(DABCYL)-NH₂
Proteins and Peptides
Sequence: Ac-Asp-Glu-Asp(EDANS)-Glu-Glu-Abu-L-lactoyl-Ser-Lys(DABCYL)-NH₂
CAS-No.: 188530-20-3
Mol.weight: 1548.61
SumFormula: C₆₈H₈₉N₁₅O₂₅S
Bulk item no.: M-2235
Synonym(s):
EINECS no.: -
MDL no.: MFCD21332883

Internally quenched fluorogenic (FRET) substrate for the continuous monitoring of HCV NS3 protease activity.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty