(4-Nitrophenyl)-β-D-galactopyranoside
Supplier: BACHEM AMERICAS INC
Synonyms:
pNPG
, 4-Nitrophenyl β-D-Galactopyranoside
4013562.0025
4013562.0005
M-1625.0025BAEA
731.51
USD
M-1625.0025BA
M-1625.0005BA
(4-Nitrophenyl)-β-D-galactopyranoside
(4-Nitrophenyl) β-D-galactopyranoside
Sequence: 4-Nitrophenyl β-D-galactopyranoside
See also the indigogenic β-galactosidase substrates M-1190 (X-Gal) and Q-1270.
Synonym(s): 4-Nitrophenyl β-D-galactoside-(1,5)
Formula:
C₁₂H₁₅NO₈ MW: 301.25 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00063256 CAS Number: 3150-24-1 EINECS: 221-584-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...