BACEM-1495

4009467.0005 4009467.0025
M-1495.0005BAEA 557.21 USD
M-1495.0005BA M-1495.0025BA
BACEM-1495
Sequence: O-Hippuryl-L-β-phenyllactic acid · sodium salt
Synonym(s): O-(Bz-Gly)-L-β-phenyllactic acid · sodium salt

Substrate for kinetic studies with carboxypeptidase A and carboxypeptidase Y. CAS-Number (net) 3675-74-9.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR